Phthalimide potassium salt ≥98%

Supplier: BeanTown Chemical

123205-100G 123205-2.5KG 123205-500G
BT123205-100GEA 37.18 USD
BT123205-100G BT123205-2.5KG BT123205-500G
Phthalimide potassium salt ≥98%
Phthalimide potassium salt

CAS: 1074-82-4; EC No: 214-046-6; MDL No: MFCD00005887 Powder; Molecular Formula: C8H4KNO2; MW: 185.23 Melting Point: 300° Density (g/mL): 1.63 Moisture Sensitive


  • Moisture Sensitive
  • Powder

Formula: C₈H₄KNO₂
Melting Pt: >300 °C
Density: 1.63 g/mL
MDL Number: MFCD00005887
CAS Number: 1074-82-4
EINECS: 214-046-6

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR