4-Chloroquinoline ≥99%
Supplier: BeanTown Chemical
123195-1G
123195-5G
BT123195-1GEA
99
USD
BT123195-1G
BT123195-5G
4-Chloroquinoline ≥99%
4-Chloroquinoline
CAS: 611-35-8; EC No: 210-267-7; MDL No: MFCD00006773
Liquid/Low Melting Solid; Molecular Formula: C9H6ClN ; MW: 163.60
Melting Point: 28-31°; Boiling Point: 260-261°; Flash point:
- Liquid/Low Melting Solid
Formula:
C₉H₆ClN Boiling Pt: 260…261 °C Melting Pt: 28…31 °C Density: 1.25 g/mL Flash Pt: >110 °C (>230 °F) |
MDL Number:
MFCD00006773 CAS Number: 611-35-8 EINECS: 210-267-7 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...