7-Quinolinol ≥98%
Supplier: BeanTown Chemical
123190-1G
123190-5G
BT123190-1GEA
49
USD
BT123190-1G
BT123190-5G
7-Quinolinol ≥98%
7-Quinolinol
CAS: 580-20-1; EC No: 209-457-2; MDL No:
MFCD00016730; RTECS: VC4160000
Solid; Molecular Formula: C9H7NO; MW: 145.16
Melting Point: 239-244°
- RTECS: VC4160000
- Solid
Formula:
C₉H₇NO Melting Pt: 239…244 °C |
MDL Number:
MFCD00016730 CAS Number: 580-20-1 EINECS: 209-457-2 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...