Bis(2-ethylhexyl)phthalate ≥98%
Supplier: BeanTown Chemical
Synonyms:
Di-iso-octyl phthalate, Dioctyl phthalate, Di-(2-ethylhexyl) phthalate, DEHP, Di(2-ethylhexyl) phthalate, Phthalic acid bis(2-ethylhexyl) ester
123150-2.5KG
123150-500G
BT123150-2.5KGEA
78.73
USD
BT123150-2.5KG
BT123150-500G
Bis(2-ethylhexyl)phthalate ≥98%
Bis(2-ethylhexyl)phthalate
CAS: 117-81-7; EC No: 204-211-0; MDL No: MFCD00009493; RTECS: TI0350000
Liquid; Molecular Formula: C24H38O4; MW: 390.56
Melting Point: -50°; Boiling Point: 384°; Flash point: 207°C (405°F)
Density (g/mL): 0.985; Refractive Index: 1.486
- RTECS: TI0350000
- Refractive index: 1.486
- Liquid
Formula:
C₂₄H₃₈O₄ MW: 390.56 g/mol Boiling Pt: 384 °C Melting Pt: –50 °C Density: 0.985 g/mL Flash Pt: 207 °C (405 °F) |
MDL Number:
MFCD00009493 CAS Number: 117-81-7 EINECS: 204-211-0 UN: 3082 ADR: 9,III |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...