2-Aminobenzenesulfonamide ≥98%
Supplier: BeanTown Chemical
123140-100G
123140-25G
123140-5G
BT123140-100GEA
1105
USD
BT123140-100G
BT123140-25G
BT123140-5G
2-Aminobenzenesulfonamide ≥98%
2-Aminobenzenesulphonamide
CAS: 3306-62-5; EC No: 221-988-1; MDL No: MFCD00007932; RTECS: DA9449000
Solid; Molecular Formula: C6H8N2O2S; MW: 172.21
Melting Point: 155-157°
- RTECS: DA9449000
- Solid
Formula:
C₆H₈N₂O₂S MW: 172.21 g/mol Melting Pt: 155…157 °C |
MDL Number:
MFCD00007932 CAS Number: 3306-62-5 EINECS: 221-988-1 |
Specification Test Results
Assay | 98% |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...