Copper(I) telluride ≥99.5%
Supplier: BeanTown Chemical
123130-25G
123130-5G
BT123130-25GEA
195
USD
BT123130-25G
BT123130-5G
Copper(I) telluride ≥99.5%
Copper(I) telluride
CAS: 12019-52-2; EC No: 234-646-1; MDL No: MFCD00049727
3mm & Down Pieces; Molecular Formula: Cu2Te; MW: 254.68
- 3mm & Down Pieces
Formula:
Cu₂Te MW: 254.69 Melting Pt: 1125 Density: 4.6 |
MDL Number:
MFCD00049727 CAS Number: 12019-52-2 EINECS: 234-646-1 |
Specification Test Results
Assay | 99.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...