Gallium phosphide ≥99.999% (trace metals basis)

Supplier: BeanTown Chemical

123125-1G 123125-5G
BT123125-1GEA 60 USD
BT123125-1G BT123125-5G
Gallium phosphide ≥99.999% (trace metals basis)
Gallium phosphide

CAS: 12063-98-8; EC No: 235-057-2; MDL No: MFCD00016109 6mm & Down Pieces; Molecular Formula: GaP; MW: 100.69


  • 6mm & Down Pieces

Formula: GaP
MDL Number: MFCD00016109
CAS Number: 12063-98-8
EINECS: 235-057-2

Order Now

Specification Test Results

Assay 99.999%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR