Isosorbide ≥98%
Supplier: BeanTown Chemical
123105-1KG
123105-250G
123105-50G
BT123105-1KGEA
304
USD
BT123105-1KG
BT123105-250G
BT123105-50G
Isosorbide ≥98%
Isosorbide
CAS: 652-67-5; EC No: 211-492-3; MDL No: MFCD00064827; RTECS: LZ4380000
Flakes; Molecular Formula: C6H10O4; MW: 146.14
Melting Point: 60-63°
Optical Rotation: [α]20/D +45.2°, c = 3 in water
Hygroscopic
- RTECS: LZ4380000
- Optical rotation: [α]20/D +45.2°, c = 3 in water
- Hygroscopic
- Flakes
Formula:
C₆H₁₀O₄ Melting Pt: 60…63 °C |
MDL Number:
MFCD00064827 CAS Number: 652-67-5 EINECS: 211-492-3 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...