cis-1,2-Cyclohexanedicarboxylic acid ≥98%

Supplier: BeanTown Chemical

Synonyms: cis-Cyclohexane-1,2-dicarboxylic acid

123095-100G 123095-25G
BT123095-100GEA 141 USD
BT123095-100G BT123095-25G
cis-1,2-Cyclohexanedicarboxylic acid ≥98%
cis-1,2-Cyclohexanedicarboxylic acid

CAS: 610-09-3; MDL No: MFCD00064949 Solid; Molecular Formula: C8H12O4 ; MW: 172.18 Melting Point: 188-192°


  • Solid

Formula: C₈H₁₂O₄
Melting Pt: 188…192 °C
MDL Number: MFCD00064949
CAS Number: 610-09-3

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR