1-Bromonaphthalene ≥97%
Supplier: BeanTown Chemical
Synonyms:
α-Bromonaphthalene
123075-10G
123075-250G
123075-50G
BT123075-10GEA
21.9
USD
BT123075-10G
BT123075-250G
BT123075-50G
1-Bromonaphthalene ≥97%
1-Bromonaphthalene
CAS: 90-11-9; EC No: 201-965-2; MDL No: MFCD00003868; RTECS: QJ1545000
Liquid; Molecular Formula: C10H7Br; MW: 207.07
Melting Point: -2°; Boiling Point: 133-134°/10 mmHg; Flash point: 110°C (230°F)
Density (g/mL): 1.48; Refractive Index: 1.657
- RTECS: QJ1545000
- Refractive index: 1.657
- Liquid
Formula:
C₁₀H₇Br MW: 207.07 g/mol Boiling Pt: 133…134 °C/10 mmHg Melting Pt: –2 °C Density: 1.48 g/mL Flash Pt: 110 °C (230 °F) |
MDL Number:
MFCD00003868 CAS Number: 90-11-9 EINECS: 201-965-2 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...