1,1,7,7-Tetramethyl-8-hydroxyjulolidine
Supplier: BeanTown Chemical
Synonyms:
1,1,7,7-Tetramethyl-1,2,3,5,6,7-hexahydropyrido[3,2,1-ij]quinolin-8-ol
123045-1G
123045-5G
BT123045-1GEA
162
USD
BT123045-1G
BT123045-5G
1,1,7,7-Tetramethyl-8-hydroxyjulolidine
1,1,7,7-Tetramethyl-8-hydroxyjulolidine
CAS: 115704-83-1
Solid; Molecular Formula: C16H23NO; MW: 245.36
- Solid
Formula:
C₁₆H₂₃NO MW: 245.366 g/mol |
MDL Number:
MFCD09264711 CAS Number: 115704-83-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...