Xylene cyanole FF ≥70% (dye content)
Supplier: BeanTown Chemical
Synonyms:
C.I. 42135
122985-10G
122985-50G
BT122985-10GEA
42.57
USD
BT122985-10G
BT122985-50G
Xylene cyanole FF ≥70% (dye content)
Xylene cyanole FF
CAS: 2650-17-1; EC No: 220-167-5; MDL No: MFCD00019481
Crystalline; Molecular Formula: C25H27N2NaO6S2 ; MW: 538.61
Melting Point: 295° (decomposes)
- Crystalline
Formula:
C₂₅H₂₇N₂NaO₆S₂ MW: 538.62 g/mol Melting Pt: 295 °C (dec.) |
MDL Number:
MFCD00040657 CAS Number: 2650-17-1 EINECS: 220-167-5 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...