Quinaldic acid ≥98%
Supplier: BeanTown Chemical
122980-100G
122980-25G
122980-5G
BT122980-100GEA
887
USD
BT122980-100G
BT122980-25G
BT122980-5G
Quinaldic acid ≥98%
Quinaldic acid
CAS: 93-10-7; EC No: 202-218-3; MDL No: MFCD00006752; RTECS: UZ9100000
Crystalline; Molecular Formula: C10H7NO2; MW: 173.17
Melting Point: 156-158°
- RTECS: UZ9100000
- Crystalline
Formula:
C₁₀H₇NO₂ Melting Pt: 156…158 °C |
MDL Number:
MFCD00006752 CAS Number: 93-10-7 EINECS: 202-218-3 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...