Calcium chloride, anhydrous ≥97%

Supplier: BeanTown Chemical

Synonyms: Chloro calcium, Calcium dichloride, CaCl2

122950-1KG 122950-250G 122950-5KG
BT122950-1KGEA 75.03 USD
BT122950-1KG BT122950-250G BT122950-5KG
Calcium chloride, anhydrous ≥97%
Calcium chloride

CAS: 10043-52-4; EC No: 233-140-8; MDL No: MFCD00010903; RTECS: EV9800000 Beads/Powder; Molecular Formula: CaCl2; MW: 110.98 Melting Point: 772° Hygroscopic


  • RTECS: EV9800000
  • Hygroscopic
  • Beads/Powder

Formula: CaCl₂
MW: 110.98 g/mol
Boiling Pt: 1600
Melting Pt: 772 °C
MDL Number: MFCD00010903
CAS Number: 10043-52-4
EINECS: 233-140-8
Merck Index: 14,01659

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR