Murexide

Supplier: BeanTown Chemical

Synonyms: Ammonium purpurate, Murexide triturate, Ammonium 5-(2,4,6-trioxoperhydropyrimidin-5-ylideneamino)barbiturate, Purpuric acid ammonium salt

122910-10G 122910-250G 122910-50G
BT122910-10GEA 55.75 USD
BT122910-10G BT122910-250G BT122910-50G
Murexide
Murexide

CAS: 3051-09-0; EC No: 221-266-6; MDL No: MFCD00012777 Powder; Molecular Formula: C8H8N6O6; MW: 284.19 Melting Point: 300°


  • Powder

Formula: C₈H₈N₆O₆
Melting Pt: >300 °C
MDL Number: MFCD00012777
CAS Number: 3051-09-0
EINECS: 221-266-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR