4-Cyano-3-fluorophenyl-trans-4-(4-ethylcyclohexyl)benzoate ≥97%

Supplier: BeanTown Chemical

Synonyms: 4-Cyano-3-fluorophenyl 4-(trans-4-butylcyclohexyl)benzoate, 4-Cyano-3-fluorophenyl 4-(trans-4-butylcyclohexyl)-benzoate

122900-1G 122900-5G
BT122900-1GEA 103 USD
BT122900-1G BT122900-5G
4-Cyano-3-fluorophenyl-trans-4-(4-ethylcyclohexyl)benzoate ≥97%
4-Cyano-3-fluorophenyl trans-4-(4-n-butylcyclohexyl)benzoate

CAS: 92118-83-7; MDL No: MFCD11053366 Solid; Molecular Formula: C24H26FNO2; MW: 379.47 Melting Point: 79-81°


  • Solid

Formula: C₂₄H₂₆FNO₂
MW: 379.475 g/mol
Melting Pt: 79…81 °C
MDL Number: MFCD11053366
CAS Number: 92118-83-7

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR