4-Aminobenzamide ≥98%
Supplier: BeanTown Chemical
122885-100G
122885-25G
122885-500G
BT122885-100GEA
66
USD
BT122885-100G
BT122885-25G
BT122885-500G
4-Aminobenzamide ≥98%
4-Aminobenzamide
CAS: 2835-68-9; EC No: 220-612-3; MDL No: MFCD00007999
Solid; Molecular Formula: C7H8N2O ; MW: 136.15
Melting Point: 181-183°
- Solid
Formula:
C₇H₈N₂O Melting Pt: 181…183 °C |
MDL Number:
MFCD00007999 CAS Number: 2835-68-9 EINECS: 220-612-3 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...