Sodium dichloroacetate ≥97%

Supplier: BeanTown Chemical

Synonyms: Sodium 2,2-dichloroacetate

122870-10G 122870-250G 122870-50G
BT122870-10GEA 88.47 USD
BT122870-10G BT122870-250G BT122870-50G
Sodium dichloroacetate ≥97%
Sodium dichloroacetate

CAS: 2156-56-1; EC No: 218-461-3; MDL No: MFCD00070489 Solid; Linear Formula: Cl2CHCOONa; MW: 150.93 Melting Point: 198° (decomposes) Moisture Sensitive


  • Moisture Sensitive
  • Solid

Formula: C₂HCl₂NaO₂
MW: 150.92 g/mol
Melting Pt: 198 °C (dec.)
MDL Number: MFCD00070489
CAS Number: 2156-56-1
EINECS: 218-461-3

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR