4-(Aminomethyl)benzoic acid ≥97%

Supplier: BeanTown Chemical

122860-100G 122860-25G 122860-500G
BT122860-100GEA 184 USD
BT122860-100G BT122860-25G BT122860-500G
4-(Aminomethyl)benzoic acid ≥97%
4-(Aminomethyl)benzoic acid

CAS: 56-91-7; EC No: 200-297-9; MDL No: MFCD00010203 Crystalline/Powder; Molecular Formula: C8H9NO2; MW: 151.16 Melting Point: 300°


  • Crystalline/Powder

Formula: C₈H₉NO₂
MW: 151.16 g/mol
Melting Pt: >300 °C
MDL Number: MFCD00010203
CAS Number: 56-91-7
EINECS: 200-297-9

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR