Formaldehyde sodium bisulfite ≥95%

Supplier: BeanTown Chemical

Synonyms: Sodium Hydroxymethanesulphonate, Hydroxymethanesulphonic acid sodium salt

122855-100G 122855-2.5KG 122855-500G
BT122855-100GEA 34 USD
BT122855-100G BT122855-2.5KG BT122855-500G
Formaldehyde sodium bisulfite ≥95%
Formaldehyde sodium bisulfite

CAS: 870-72-4; EC No: 212-800-9; MDL No: MFCD00044664; RTECS: PB2200000 Solid; Linear Formula: HOCH2SO3Na; MW: 134.09 Melting Point: 200° (decomposes) Hygroscopic


  • RTECS: PB2200000
  • Hygroscopic
  • Solid

Formula: CH₄O₄S
MW: 134.08 g/mol
Melting Pt: 200 °C (dec.)
MDL Number: MFCD00044664
CAS Number: 870-72-4
EINECS: 212-800-9

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR