Ammonium acetate ≥97.0% ACS

Supplier: BeanTown Chemical

122845-2KG 122845-500G
BT122845-2KGEA 181.5 USD
BT122845-2KG BT122845-500G
Ammonium acetate ≥97.0% ACS
Ammonium acetate

CAS: 631-61-8; EC No: 211-162-9; MDL No: MFCD00013066; RTECS: AF3675000 Crystalline; Linear Formula: CH3CO2NH4; MW: 77.08 Melting Point: 110-112° (decomposes) Hygroscopic


  • RTECS: AF3675000
  • Hygroscopic
  • Crystalline

Formula: H₃CCOONH₄
Melting Pt: 110…112 °C (dec.)
MDL Number: MFCD00013066
CAS Number: 631-61-8
EINECS: 211-162-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd160345483553318b94053de396335a9b18425f&vid=c6375b539008d6b43f50408d30b61261444faebc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 97.0% min.
Insoluble matter ≤0.005%
pH of 5% solution at 25°C 6.7-7.0
Chloride (Cl) ≤5ppm
Nitrate (NO3) ≤0.001%
Sulfate (SO4) ≤0.001%
Heavy metals (as Pb) ≤5ppm
Iron (Fe) ≤5ppm
Residue on ignition ≤0.005%
Appearance Crystals
Solubility Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR