Ammonium dimolybdate

Supplier: BeanTown Chemical

122805-1KG 122805-250G 122805-50G
BT122805-1KGEA 196 USD
BT122805-1KG BT122805-250G BT122805-50G
Ammonium dimolybdate
Ammonium dimolybdate

CAS: 27546-07-2; EC No: 248-517-2; MDL No: MFCD00010882 Powder; Linear Formula: (NH4)2Mo2O7 ; MW: 339.95


  • Powder

Formula: H₈Mo₂N₂O₇
MDL Number: MFCD00010882
CAS Number: 27546-07-2
EINECS: 248-517-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR