1-Benzyl-4-piperidone ≥98%
Supplier: BeanTown Chemical
122790-100G
122790-25G
122790-500G
BT122790-100GEA
112
USD
BT122790-100G
BT122790-25G
BT122790-500G
1-Benzyl-4-piperidone ≥98%
1-Benzyl-4-piperidone
CAS: 3612-20-2; EC No: 222-782-4; MDL No: MFCD00006192
Liquid; Molecular Formula: C12H15NO; MW: 189.25
Boiling Point: 248°; Flash point: 71°C (160°F)
Density (g/mL): 1.021; Refractive Index: 1.541
- Refractive index: 1.541
- Liquid
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...