2,3-Diamino-6-methoxypyridine dihydrochloride ≥99%

Supplier: BeanTown Chemical

Synonyms: 6-Methoxypyridine-2,3-diamine dihydrochloride

122780-1G 122780-250MG 122780-5G
BT122780-1GEA 114 USD
BT122780-1G BT122780-250MG BT122780-5G
2,3-Diamino-6-methoxypyridine dihydrochloride ≥99%
2,3-Diamino-6-methoxypyridine dihydrochloride

CAS: 94166-62-8; EC No: 303-358-9; MDL No: MFCD07787253 Powder; Molecular Formula: C6H9N3O·2HCl; MW: 212.08


  • Powder

Formula: C₆H₁₁Cl₂N₃O
MW: 212.074 g/mol
MDL Number: MFCD00137799
CAS Number: 94166-62-8
EINECS: 303-358-9

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR