Benzamidine hydrochloride hydrate, (H₂O 10-14%) ≥98%
Supplier: BeanTown Chemical
122775-100G
122775-25G
122775-5G
BT122775-100GEA
181
USD
BT122775-100G
BT122775-25G
BT122775-5G
Benzamidine hydrochloride hydrate, (H₂O 10-14%) ≥98%
Benzamidine hydrochloride hydrate
CAS: 206752-36-5; EC No: 216-795-4; MDL No: MFCD00066285; RTECS: CV6260000
Crystalline/Powder; Molecular Formula: C7H8N26·HCl·xH2O; MW: 156.61 (anhydrous)
Melting Point: 86-88°
Hygroscopic
- RTECS: CV6260000
- Hygroscopic
- Crystalline/Powder
Formula:
C₆H₅C(=NH)NH₂·HCl·H₂O MW: 174.63 g/mol Melting Pt: 86…88 °C |
MDL Number:
MFCD00066285 CAS Number: 206752-36-5 EINECS: 216-795-4 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...