tert-Butyl-N-hydroxycarbamate ≥98%
Supplier: BeanTown Chemical
Synonyms:
N-Boc-hydroxylamine, N-tert-Butoxycarbonylhydroxylamine, N-Tert-Boc-Hydroxylamine
122755-1G
122755-25G
122755-5G
BT122755-1GEA
26
USD
BT122755-1G
BT122755-25G
BT122755-5G
tert-Butyl-N-hydroxycarbamate ≥98%
tert-Butyl-N-hydroxycarbamate
CAS: 36016-38-3; EC No: 252-836-2; MDL No: MFCD00002107
Crystalline/Powder; Linear Formula: (CH3)3COCONHOH; MW: 133.15
Melting Point: 53-55°
Moisture Sensitive
- Moisture Sensitive
- Crystalline/Powder
Formula:
C₅H₁₁NO₃ MW: 133.15 g/mol Melting Pt: 53…55 °C |
MDL Number:
MFCD00002107 CAS Number: 36016-38-3 EINECS: 252-836-2 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...