Trimethylphosphine oxide

Supplier: BeanTown Chemical

Synonyms: Trimethylphosphineoxide

122705-1G 122705-5G
BT122705-1GEA 122.14 USD
BT122705-1G BT122705-5G
Trimethylphosphine oxide
Trimethylphosphine oxide

CAS: 676-96-0; EC No: 211-633-9; MDL No: MFCD00013910 Crystalline; Linear Formula: (CH3)3P(O); MW: 92.08 Melting Point: 140-141°


  • Crystalline

Formula: C₃H₉OP
MW: 92.08 g/mol
Melting Pt: 140…141 °C
MDL Number: MFCD00013910
CAS Number: 676-96-0
EINECS: 211-633-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR