Manganese(IV) oxide ≥99.995%
Supplier: BeanTown Chemical
Synonyms:
Manganese dioxide, Manganese(IV) oxide, activated, Manganese peroxide, Mangan(IV)-oxid
122685-100G
122685-25G
122685-5G
BT122685-100GEA
487
USD
BT122685-100G
BT122685-25G
BT122685-5G
Manganese(IV) oxide ≥99.995%
Manganese(IV) oxide
CAS: 1313-13-9; EC No: 215-202-6; MDL No: MFCD00003463; RTECS: OP0350000
Powder; Molecular Formula: MnO2; MW: 86.94
Melting Point: 535° (decomposes)
Density (g/mL): 5.026
- RTECS: OP0350000
- Powder
Formula:
MnO₂ MW: 86.94 g/mol Melting Pt: 535 (dec.) Density: 5.026 g/mL |
MDL Number:
MFCD00003463 CAS Number: 1313-13-9 EINECS: 215-202-6 Merck Index: 14,05730 |
Specification Test Results
Assay | 99.995% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...