3,4,5-Tribromo-1H-pyrazole ≥97%

Supplier: BeanTown Chemical

Synonyms: 3,4,5-Tribromopyrazole

122675-100G 122675-25G 122675-5G
BT122675-100GEA 1079 USD
BT122675-100G BT122675-25G BT122675-5G
3,4,5-Tribromo-1H-pyrazole ≥97%
3,4,5-Tribromo-1H-pyrazole

CAS: 17635-44-8; MDL No: MFCD00040248 Solid; Molecular Formula: C3HBr3N2; MW: 304.77


  • Solid

Formula: C₃HBr₃N₂
MW: 304.78 g/mol
MDL Number: MFCD00040248
CAS Number: 17635-44-8

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR