Bismuth ≥99.999% (trace metals basis), foil, Thickness: 2.0 mm

Supplier: BeanTown Chemical

122640-25X25MM 122640-50X50MM
BT122640-25X25EA 786.65 USD
BT122640-25X25 BT122640-50X50
Bismuth ≥99.999% (trace metals basis), foil, Thickness: 2.0 mm
Bismuth

CAS: 7440-69-9; EC No: 231-177-4; MDL No: MFCD00134033 Solid; Molecular Formula: Bi; MW: 208.98 Melting Point: 271°; Boiling Point: 1560° Density (g/mL): 9.8


  • Solid

Formula: Bi
Boiling Pt: 1560 °C
Melting Pt: 271 °C
Density: 9.8 g/mL
MDL Number: MFCD00134033
CAS Number: 7440-69-9
EINECS: 231-177-4

Order Now

Specification Test Results

Assay 99.999%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR