1,3-Diphenylurea ≥98%

Supplier: BeanTown Chemical

Synonyms: N,N’-Diphenylurea, N,N'-Diphenylurea

122610-100G 122610-25G
BT122610-100GEA 114 USD
BT122610-100G BT122610-25G
1,3-Diphenylurea ≥98%
1,3-Diphenylurea

CAS: 102-07-8; EC No: 203-003-7; MDL No: MFCD00003017; RTECS: FD9800000 Crystalline; Linear Formula: (C6H5NH)2CO; MW: 212.25 Melting Point: 239-241°; Boiling Point: 262° Density (g/mL): 1.239


  • RTECS: FD9800000
  • Crystalline

Formula: C₁₃H₁₂N₂O
MW: 212.25 g/mol
Boiling Pt: 262 °C
Melting Pt: 239…241 °C
Density: 1.239 g/mL
MDL Number: MFCD00003017
CAS Number: 102-07-8
EINECS: 203-003-7

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR