2-Chloro-N-(2,6-diethylphenyl)acetamide ≥96%

Supplier: BeanTown Chemical

Synonyms: 2-Chloro-2',6'-diethylacetanilide

122605-25G 122605-5G
BT122605-25GEA 253 USD
BT122605-25G BT122605-5G
2-Chloro-N-(2,6-diethylphenyl)acetamide ≥96%
2-Chloro-N-(2,6-diethylphenyl)acetamide

CAS: 6967-29-9; EC No: 214-460-7; MDL No: MFCD00124800 Powder; Molecular Formula: C12H16ClNO ; MW: 225.71


  • Powder

Formula: C₁₂H₁₆ClNO
MW: 225.72 g/mol
MDL Number: MFCD00124800
CAS Number: 6967-29-9
EINECS: 214-460-7

Order Now

Specification Test Results

Assay 96%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR