N-(Diphenylmethylene)glycine ethyl ester ≥98%
Supplier: BeanTown Chemical
122590-25G
122590-5G
BT122590-25GEA
242
USD
BT122590-25G
BT122590-5G
N-(Diphenylmethylene)glycine ethyl ester ≥98%
N-(Diphenylmethylene)glycine ethyl ester
CAS: 69555-14-2; MDL No: MFCD00010590
Crystalline; Linear Formula: (C6H5)2C=NCH2COOCH2CH3; Molecular Formula: C17H17NO2 ; MW: 267.32
Melting Point: 51-53°; Flash point: 110°C (230°F)
Moisture Sensitive
- Moisture Sensitive
- Crystalline
Formula:
C₁₇H₁₇NO₂ Melting Pt: 51…53 °C Flash Pt: 110 °C (230 °F) |
MDL Number:
MFCD00010590 CAS Number: 69555-14-2 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...