2-Methoxynicotinic acid ≥98%
Supplier: BeanTown Chemical
Synonyms:
2-Methoxy-3-pyridinecarboxylic acid, 2-Methoxypyridine-3-carboxylic acid
122575-1G
122575-5G
BT122575-1GEA
51
USD
BT122575-1G
BT122575-5G
2-Methoxynicotinic acid ≥98%
2-Methoxynicotinic acid
CAS: 16498-81-0; MDL No: MFCD00085110
Solid; Molecular Formula: C7H6NO3; MW: 153.14
Melting Point: 146-150°
- Solid
Formula:
C₇H₇NO₃ Melting Pt: 146…150 °C |
MDL Number:
MFCD00085110 CAS Number: 16498-81-0 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...