Aluminium oxide, γ-phase ≥99%

Supplier: BeanTown Chemical

122565-100G 122565-25G 122565-500G
BT122565-100GEA 171 USD
BT122565-100G BT122565-25G BT122565-500G
Aluminium oxide, γ-phase ≥99%
Aluminium oxide, γ-phase

CAS: 1344-28-1; EC No: 215-691-6; MDL No: MFCD00003424 20nm APS Powder, S.A. 100-200 m2/g; Molecular Formula: Al2O3; MW: 101.96 Melting Point: 2040°


  • 20nm APS Powder, S.A. 100-200 m²/g

Formula: Al₂O₃
MW: 101.96 g/mol
Melting Pt: 2040 °C
MDL Number: MFCD00003424
CAS Number: 1344-28-1
EINECS: 215-691-6

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR