2,4-Dihydroxybenzaldehyde ≥98%

Supplier: BeanTown Chemical

122555-100G 122555-25G 122555-500G
BT122555-100GEA 200 USD
BT122555-100G BT122555-25G BT122555-500G
2,4-Dihydroxybenzaldehyde ≥98%
2,4-Dihydroxybenzaldehyde

CAS: 95-01-2; EC No: 202-383-1; MDL No: MFCD00011686; RTECS: VH3600000 Solid; Molecular Formula: C7H6O3; MW: 138.12 Melting Point: 135-137°; Boiling Point: 220-228°/22 mmHg Air Sensitive


  • RTECS: VH3600000
  • Air Sensitive
  • Solid

Formula: C₇H₆O₃
MW: 138.12 g/mol
Boiling Pt: 220…228 °C/22 mmHg
Melting Pt: 135…137 °C
MDL Number: MFCD00011686
CAS Number: 95-01-2
EINECS: 202-383-1

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR