Ethyl-3-aminopropanoate hydrochloride ≥98%

Supplier: BeanTown Chemical

Synonyms: beta-Alanine ethyl ester hydrochloride, H-beta-Ala-OEt.HCl, Ethyl 3-Aminopropionate Hydrochloride, β-Alanine ethyl ester hydrochloride, 3-Aminopropionic Acid Ethyl Ester Hydrochloride, Ethyl 3-aminopropanoate hydrochloride, 3-Aminopropionic acid Ethyl Ester Hydrochloride, H-β-Ala-OEt·HCl, Ethyl 3-aminopropionate HCl

122540-10G 122540-250G 122540-50G
BT122540-10GEA 51 USD
BT122540-10G BT122540-250G BT122540-50G
Ethyl-3-aminopropanoate hydrochloride ≥98%
Ethyl-3-aminopropanoate hydrochloride

CAS: 4244-84-2; EC No: 224-203-0; MDL No: MFCD00012909 Crystalline; Linear Formula: NH2CH2CH2COOCH2CH3·HCl; Molecular Formula: C5H11NO2·HCl; MW: 153.61 Melting Point: 67-72° Hygroscopic


  • Hygroscopic
  • Crystalline

Formula: C₅H₁₁NO₂·HCl
MW: 153.6 g/mol
Melting Pt: 67…72 °C
MDL Number: MFCD00012909
CAS Number: 4244-84-2
EINECS: 224-203-0

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR