(±)-Aspartic acid ≥98%
Supplier: BeanTown Chemical
Synonyms:
2-Aminobutanedioic acid, DL-Aspartic acid
122535-100G
122535-2.5KG
122535-500G
BT122535-100GEA
20
USD
BT122535-100G
BT122535-2.5KG
BT122535-500G
(±)-Aspartic acid ≥98%
(±)-Aspartic acid
CAS: 617-45-8; EC No: 217-234-6; MDL No: MFCD00063083; RTECS: CI9097800
Solid; Linear Formula: HOOCCH2CH(NH2)COOH; Molecular Formula: C4H6NO4; MW: 133.10
Melting Point:
- RTECS: CI9097800
- Solid
Formula:
C₄H₇NO₄ Melting Pt: >300 °C |
MDL Number:
MFCD00063083 CAS Number: 617-45-8 EINECS: 217-234-6 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...