Tris(2-methylphenyl)phosphine ≥98%
Supplier: BeanTown Chemical
122525-1G
122525-25G
122525-5G
BT122525-1GEA
31.28
USD
BT122525-1G
BT122525-25G
BT122525-5G
Tris(2-methylphenyl)phosphine ≥98%
Tris(2-methylphenyl)phosphine
CAS: 6163-58-2; EC No: 228-193-9; MDL No: MFCD00008514
Crystalline/Powder; Linear Formula: (CH3C6H4)3P; Molecular Formula: C21H21P; MW: 304.37
Melting Point: 123-125°
Light Sensitive
- Light Sensitive
- Crystalline/Powder
Formula:
C₂₁H₂₁P Melting Pt: 123…125 °C |
MDL Number:
MFCD00008514 CAS Number: 6163-58-2 EINECS: 228-193-9 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...