5-Amino-1H-tetrazole ≥98%

Supplier: BeanTown Chemical

Synonyms: 1H-Tetrazol-5-ylamine

122520-100G 122520-25G
BT122520-100GEA 100.66 USD
BT122520-100G BT122520-25G
5-Amino-1H-tetrazole ≥98%
5-Amino-1H-tetrazole

CAS: 4418-61-5; EC No: 224-581-7; MDL No: MFCD00005248; RTECS: XF7465000 Solid; Molecular Formula: CH3N5; MW: 85.07 Melting Point: 200-206°


  • RTECS: XF7465000
  • Solid

Formula: CH₃N₅
MW: 85.07 g/mol
Melting Pt: 200…206 °C
MDL Number: MFCD00005248
CAS Number: 4418-61-5
EINECS: 224-581-7
UN: 1325
ADR: 4.1,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR