3,5-Di-tert-butyl-4-hydroxybenzaldehyde ≥98%
Supplier: BeanTown Chemical
122515-100G
122515-25G
122515-5G
BT122515-100GEA
247
USD
BT122515-100G
BT122515-25G
BT122515-5G
3,5-Di-tert-butyl-4-hydroxybenzaldehyde ≥98%
3,5-Di-tert-butyl-4-hydroxybenzaldehyde
CAS: 1620-98-0; EC No: 216-592-0; MDL No: MFCD00008826; RTECS: CU5610070
Crystalline; Molecular Formula: C15H22O2; MW: 234.33
Air Sensitive
- RTECS: CU5610070
- Air Sensitive
- Crystalline
Formula:
C₁₅H₂₂O₂ |
MDL Number:
MFCD00008826 CAS Number: 1620-98-0 EINECS: 216-592-0 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...