Silver(I) citrate hydrate

Supplier: BeanTown Chemical

122490-100G 122490-25G 122490-5G
BT122490-100GEA 343 USD
BT122490-100G BT122490-25G BT122490-5G
Silver(I) citrate hydrate
Silver(I) citrate hydrate

CAS: 206986-90-5; MDL No: MFCD00150589 Powder; Linear Formula: AgO2CCH2C(OH)(CO2Ag)CH2CO2Ag·xH2O; Molecular Formula: Ag3C6H5O7·xH2O; MW: 512.70 (anhydrous) Melting Point: 170° (decomposes) Light Sensitive


  • Light Sensitive
  • Powder

Formula: C₆H₅Ag₃O₇·xH₂O
MDL Number: MFCD00150589
CAS Number: 206986-90-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR