Palladium ≥99.9% (trace metals basis), foil, Thickness 0.025 mm
Supplier: BeanTown Chemical
122445-25X25MM
122445-50X50MM
BT122445-25X25EA
133
USD
BT122445-25X25
BT122445-50X50
Palladium ≥99.9% (trace metals basis), foil, Thickness 0.025 mm
Palladium
CAS: 7440-05-3; EC No: 231-115-6; MDL No: MFCD00011167; RTECS: RT3480500
Solid; Molecular Formula: Pd; MW: 106.42
Melting Point: 1554°; Boiling Point: 2970°
Density (g/mL): 12.02
- RTECS: RT3480500
- Solid
Formula:
Pd MW: 106.420 g/mol Boiling Pt: 2970 °C Melting Pt: 1554 °C Density: 12.02 g/mL |
MDL Number:
MFCD00011167 CAS Number: 7440-05-3 EINECS: 231-115-6 |
Specification Test Results
Assay | 99.9% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...