Colophony (Rosin gum)
Supplier: BeanTown Chemical
122435-1KG
122435-250G
BT122435-1KGEA
64.29
USD
BT122435-1KG
BT122435-250G
Colophony (Rosin gum)
Colophony (Rosin gum)
CAS: 8050-09-7; EC No: 232-475-7; MDL No: MFCD00132205
Powder/Solid
Melting Point: 67-93°; Flash point: 188°C (370°F)
Density (g/mL): 1.304
- Powder/Solid
Melting Pt:
67…93 °C Density: 1.304 g/mL Flash Pt: 188 °C (370 °F) |
MDL Number:
MFCD00132205 CAS Number: 8050-09-7 EINECS: 232-475-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...