2,3-Dihydroxybenzaldehyde ≥97%

Supplier: BeanTown Chemical

122415-100G 122415-25G 122415-5G
BT122415-100GEA 650 USD
BT122415-100G BT122415-25G BT122415-5G
2,3-Dihydroxybenzaldehyde ≥97%
2,3-Dihydroxybenzaldehyde

CAS: 24677-78-9; EC No: 246-398-1; MDL No: MFCD00003324 Powder; Molecular Formula: C7H6O3; MW: 138.12 Melting Point: 104-108° Air Sensitive


  • Air Sensitive
  • Powder

Formula: C₇H₆O₃
MW: 138.12 g/mol
Melting Pt: 104…108 °C
MDL Number: MFCD00003324
CAS Number: 24677-78-9
EINECS: 246-398-1

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR