D-(-)-Ribose ≥98%

Supplier: BeanTown Chemical

122395-10G 122395-250G 122395-50G
BT122395-10GEA 25 USD
BT122395-10G BT122395-250G BT122395-50G
D-(-)-Ribose ≥98%
D(-)-Ribose

CAS: 50-69-1; EC No: 200-059-4; MDL No: MFCD00135453; RTECS: VJ2275000 Powder; Molecular Formula: C5H10O5; MW: 150.13 Melting Point: 88-92° Optical Rotation: [α]20/D -19.7°, c = 4 in water Hygroscopic


  • RTECS: VJ2275000
  • Optical rotation: [α]20/D -19.7°, c = 4 in water
  • Hygroscopic
  • Powder

Formula: C₅H₁₀O₅
MW: 150.13 g/mol
Melting Pt: 88…92 °C
MDL Number: MFCD00135453
CAS Number: 50-69-1
EINECS: 200-059-4

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR