(R)-(-)-1-Phenyl-1,2-ethanediol ≥99%

Supplier: BeanTown Chemical

Synonyms: (R)-1-Phenylethane-1,2-diol, (R)-(-)-α,β-Dihydroxyethylbenzene, (R)-(-)-Phenylethane-1,2-diol, (R)-(-)-Phenyl-1,2-ethanediol

122370-1G 122370-250MG
BT122370-1GEA 75 USD
BT122370-1G BT122370-250MG
(R)-(-)-1-Phenyl-1,2-ethanediol ≥99%
(R)-(-)-1-Phenyl-1,2-ethanediol

CAS: 16355-00-3; MDL No: MFCD00064262 Crystalline; Molecular Formula: C8H10O2 ; MW: 138.16 Melting Point: 64-67°; Boiling Point: 272-274° Optical Rotation: [α]20/D −69°, c = 1 in chloroform


  • Optical rotation: [α]20/D −69°, c = 1 in chloroform
  • Crystalline

Formula: C₈H₁₀O₂
MW: 138.17 g/mol
Boiling Pt: 272…274 °C
Melting Pt: 64…67 °C
MDL Number: MFCD00064262
CAS Number: 16355-00-3

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR