Barium zirconate ≥99%

Supplier: BeanTown Chemical

122365-1KG 122365-250G
BT122365-1KGEA 126.24 USD
BT122365-1KG BT122365-250G
Barium zirconate ≥99%
Barium zirconate

CAS: 12009-21-1; EC No: 234-546-8; MDL No: MFCD00014189 1-2 Micron Powder; Linear Formula: BaZrO3; MW: 276.55 Density (g/mL): 5.52


  • 1-2 Micron Powder

Formula: BaO₃Zr
MW: 276.548 g/mol
Density: 5.52 g/mL
MDL Number: MFCD00014189
CAS Number: 12009-21-1
EINECS: 234-546-8

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR