O-Dianisidine dihydrochloride ≥99%
Supplier: BeanTown Chemical
122320-1G
122320-5G
BT122320-1GEA
51.44
USD
BT122320-1G
BT122320-5G
O-Dianisidine dihydrochloride ≥99%
O-Dianisidine dihydrochloride
CAS: 20325-40-0; EC No: 243-737-5; MDL No: MFCD00012488; RTECS: DD1050000
Solid; Molecular Formula: C14H16N2O2·2HCl; MW: 317.21
Melting Point: 268°
- RTECS: DD1050000
- Solid
Formula:
C₁₄H₁₈Cl₂N₂O₂ Melting Pt: 268 °C |
MDL Number:
MFCD00012488 CAS Number: 20325-40-0 EINECS: 243-737-5 |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...