Lithium chloride/potassium chloride eutectic

Supplier: BeanTown Chemical

122310-250G 122310-50G
BT122310-250GEA 469.75 USD
BT122310-250G BT122310-50G
Lithium chloride/potassium chloride eutectic
Lithium chloride/potassium chloride eutectic

CAS: 65567-96-6; MDL No: MFCD00144623 -10 Mesh Powder; Linear Formula: LiCl·KCl; MW: Melting Point: 357° Density (g/mL): 2.02 Hygroscopic


  • Hygroscopic
  • -10 Mesh Powder

Formula: Cl₂KLi
MW: 116.938 g/mol
Melting Pt: 357 °C
Density: 2.02 g/mL
MDL Number: MFCD00144623
CAS Number: 65567-96-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR